Methyl-5-hexenoate ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Methyl 5-hexenoate, 5-Hexenoic Acid Methyl Ester
H0872-25G
H0872-5G
TCIAH0872-25GEA
2560
DKK
TCIAH0872-25G
TCIAH0872-5G
Methyl-5-hexenoate ≥98.0% (by GC)
Methyl-5-hexenoate
Formel:
C₇H₁₂O₂ Kogepunkt: 157°C Flammepunkt: 46°C |
MDL Number:
MFCD00671538 CAS-nummer: 2396-80-7 UN nummer: 3272 ADR: 3,III |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...