5-Hexenenitrile ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

H0836-25ML H0836-5ML
TCIAH0836-25MLEA 3660 DKK
TCIAH0836-25ML TCIAH0836-5ML
5-Hexenenitrile ≥95.0% (by GC)
5-Hexenenitrile
Formel: C₆H₉N
Kogepunkt: 162°C
Flammepunkt: 41°C
MDL Number: MFCD00067056
CAS-nummer: 5048-19-1
UN nummer: 1992
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52adce5b89df52c9a0af805816845987b0599144&vid=52adce5b89df52c9a0af805816845987b0599144&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR