Hexadecyltrimethylammonium hexafluorophosphate ≥98.0% (by total nitrogen basis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N-hexadecyltrimethylammoniumhexafluorophosphate, N-Hexadecyltrimethylammonium hexafluorophosphate, Cetyltrimethylammonium Hexafluorophosphate

H0733-1G H0733-25G H0733-5G
TCIAH0733-1GEA 165 DKK
TCIAH0733-1G TCIAH0733-25G TCIAH0733-5G
Hexadecyltrimethylammonium hexafluorophosphate ≥98.0% (by total nitrogen basis)
Hexadecyltrimethylammonium hexafluorophosphate
Formel: C₁₉H₄₂F₆NP
MDL Number: MFCD00144894
CAS-nummer: 101079-29-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b985f45e4fe6abb80192ef80f909b7074982ba2d&vid=f6266a60725eeb5b02553b7a73df41049623c80c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR