3-Heptenoic acid ≥90.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
H0427-25ML
H0427-5ML
TCIAH0427-25MLEA
1840
DKK
TCIAH0427-25ML
TCIAH0427-5ML
3-Heptenoic acid ≥90.0% (by GC)
3-Heptenoic acid
Formel:
C₇H₁₂O₂ Kogepunkt: 228°C |
MDL Number:
MFCD00058999 CAS-nummer: 29901-85-7 EINECS: 249-943-1 UN nummer: 3265 ADR: 8,III |
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 90.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...