Heptanoic anhydride ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

H0353-25ML H0353-500ML
TCIAH0353-25MLEA 132 DKK
TCIAH0353-25ML TCIAH0353-500ML
Heptanoic anhydride ≥97.0% (by titrimetric analysis)
Heptanoic anhydride
Formel: C₁₄H₂₆O₃
Kogepunkt: 260°C
MDL Number: MFCD00009536
CAS-nummer: 626-27-7
EINECS: 210-940-5
UN nummer: 3265
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (Morpholine Method) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR