12-Hydroxystearic acid ≥80.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H0308-25G H0308-500G
TCIAH0308-25GEA 148 DKK
TCIAH0308-25G TCIAH0308-500G
12-Hydroxystearic acid ≥80.0% (by GC)
12-Hydroxystearic acid
Formel: C₁₈H₃₆O₃
Kogepunkt: 180°C
MDL Number: MFCD00004592
CAS-nummer: 106-14-9
EINECS: 203-366-1

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green flake
Purity (GC) min. 80.0 %
Melting point 75.0 to 80.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR