4-Hydroxyphenylpyruvic acid ≥95.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H0294-1G
H0294-5G
TCIAH0294-1GEA
452
DKK
TCIAH0294-1G
TCIAH0294-5G
4-Hydroxyphenylpyruvic acid ≥95.0% (by HPLC, titration analysis)
4-Hydroxyphenylpyruvic acid
Formel:
C₉H₈O₄ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00002591 CAS-nummer: 156-39-8 EINECS: 205-852-9 |
Specifikationer Testresultater
Appearance | White to Gray to Brown powder to crystaline |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...