Ethyl 2-Hydroxybutyrate ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Ethyl 2-Hydroxybutyrate, 2-Hydroxybutyric Acid Ethyl Ester, Ethyl 2-hydroxybutanoate
H0229-25G
H0229-5G
TCIAH0229-25GEA
5870
DKK
TCIAH0229-25G
TCIAH0229-5G
Ethyl 2-Hydroxybutyrate ≥95.0% (by GC)
Ethyl-2-hydroxybutyrate
Formel:
C₆H₁₂O₃ Kogepunkt: 167°C |
MDL Number:
MFCD00014407 CAS-nummer: 52089-54-0 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...