Nonyl-4-hydroxybenzoate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Hydroxybenzoic Acid Nonyl Ester, Nonylparaben

H0217-25G H0217-500G H0217-100G
TCIAH0217-25GEA 509 DKK
TCIAH0217-25G TCIAH0217-500G TCIAH0217-100G
Nonyl-4-hydroxybenzoate ≥98.0% (by HPLC, titration analysis)
Nonyl-4-hydroxybenzoate
Formel: C₁₆H₂₄O₃
MDL Number: MFCD00016483
CAS-nummer: 38713-56-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9dd2dc1886f506ab3033abdcbe609c1e17f832bd&vid=9dd2dc1886f506ab3033abdcbe609c1e17f832bd&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 41.0 to 46.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR