Hexanenitrile ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Amyl Cyanide, Capronitrile
H0102-25ML
H0102-5ML
TCIAH0102-25MLEA
796
DKK
TCIAH0102-25ML
TCIAH0102-5ML
Hexanenitrile ≥97.0% (by GC)
Hexanenitrile
Formel:
C₆H₁₁N Molekylvægt: 97.16 g/mol Kogepunkt: 160°C Flammepunkt: 43°C |
MDL Number:
MFCD00001979 CAS-nummer: 628-73-9 EINECS: 211-052-0 UN nummer: 1993 ADR: 3 III |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...