Hexadecanamide ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Palmitamide
H0067-10G
H0067-25G
TCIAH0067-10GEA
1200
DKK
TCIAH0067-10G
TCIAH0067-25G
Hexadecanamide ≥95.0% (by GC)
Hexadecanamide
Formel:
C₁₆H₃₃NO Molekylvægt: 255.45 g/mol Kogepunkt: 236°C |
MDL Number:
MFCD00025534 CAS-nummer: 629-54-9 EINECS: 211-095-5 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 103.0 to 108.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...