Glycine sulphate ≥98.0% (by total nitrogen and titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Glycine sulphate, H-Gly-OH.1/3H2SO4

G0424-5G G0424-25G
TCIAG0424-5GEA 796 DKK
TCIAG0424-5G TCIAG0424-25G
Glycine sulphate ≥98.0% (by total nitrogen and titration analysis)
Glycine sulphate
Formel: C₂H₅NO₂
Molekylvægt: 107.76 g/mol
MDL Number: MFCD00036426
CAS-nummer: 513-29-1
EINECS: 208-154-2
Merck Index: 14,04492
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1d4ac0a4d2257d1d980abd99babd19a263db365&vid=f1d4ac0a4d2257d1d980abd99babd19a263db365&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR