(+)-Griseofulvin ≥97.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

G0384-25G G0384-5G
TCIAG0384-25GEA 641 DKK
TCIAG0384-25G TCIAG0384-5G
(+)-Griseofulvin ≥97.0% (by HPLC)
(+)-Griseofulvin
Formel: C₁₇H₁₇ClO₆
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00082343
CAS-nummer: 126-07-8
EINECS: 204-767-4
Merck Index: 14,04549

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Argentmetric Titration) min. 95.0 %
Melting point 218.0 to 222.0 deg-C
Specific rotation [a]20/D +350.0 to +364.0 deg (C=1, DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR