Galantamine hydrobromide ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Galanthamine HBr, (4aS,6R,8aS)-3-Methoxy-11-methyl-5,6,9,10,11,12-hexahydro-4aH-benzo[2,3]benzofuro[4,3-cd]azepin-6-ol hydrobromide
G0293-1G
G0293-100MG
TCIAG0293-1GEA
3950
DKK
TCIAG0293-1G
TCIAG0293-100MG
Galantamine hydrobromide ≥98.0% (by HPLC, titration analysis)
Galantamine hydrobromide
Formel:
C₁₇H₂₁NO₃·HBr Smeltepunkt: 256°C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00067672 CAS-nummer: 1953-04-4 EINECS: 217-780-5 UN nummer: 2811 ADR: 6.1,II Merck Index: 14,04340 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -92.0 to -98.0 deg (C=1.4, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...