4-Guanidinobenzoic acid methanesulphonate ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
G0248-25G
G0248-5G
TCIAG0248-25GEA
1110
DKK
TCIAG0248-25G
TCIAG0248-5G
4-Guanidinobenzoic acid methanesulphonate ≥98.0% (by titrimetric analysis)
4-Guanidinobenzoic acid methanesulphonate
Formel:
C₈H₉N₃O₂ |
MDL Number:
MFCD00191442 CAS-nummer: 148720-07-4 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...