Guanine sulphate dihydrate ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Aminohypoxanthine Sulphate

G0168-1G G0168-25G
TCIAG0168-1GEA 132 DKK
TCIAG0168-1G TCIAG0168-25G
Guanine sulphate dihydrate ≥98.0% (by HPLC, total nitrogen)
Guanine sulphate dihydrate
Formel: C₅H₅N₅O
Molekylvægt: 400.33 g/mol
MDL Number: MFCD00213671
CAS-nummer: 10333-92-3
EINECS: 233-727-9

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Solubility in hot HCl almost transparency
Drying loss 7.0 to 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR