Glycyrrhetic acid ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

G0149-1G G0149-25G
TCIAG0149-1GEA 132 DKK
TCIAG0149-1G TCIAG0149-25G
Glycyrrhetic acid ≥97.0% (by titrimetric analysis)
Glycyrrhetic acid
Formel: C₃₀H₄₆O₄
MDL Number: MFCD00003706
CAS-nummer: 471-53-4
EINECS: 207-444-6
Merck Index: 14,03590

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 97.0 %
Specific rotation [a]20/D +154.0 to +170.0 deg (C=1, Methanol)
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR