Glycylsarcosine ≥97.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

G0138-1G G0138-5G
TCIAG0138-1GEA 1130 DKK
TCIAG0138-1G TCIAG0138-5G
Glycylsarcosine ≥97.0% (by HPLC)
Glycylsarcosine
Formel: C₅H₁₀N₂O₃
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00038193
CAS-nummer: 29816-01-1
EINECS: 249-875-2

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR