Isoamyl gallate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Gallic Acid Isoamyl Ester

G0012-25G G0012-500G
TCIAG0012-25GEA 468 DKK
TCIAG0012-25G TCIAG0012-500G
Isoamyl gallate ≥98.0% (by HPLC, titration analysis)
Isoamyl gallate
Formel: C₁₂H₁₆O₅
Molekylvægt: 240.26 g/mol
MDL Number: MFCD00016431
CAS-nummer: 2486-02-4
EINECS: 219-626-2

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 145.0 to 148.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR