4-Fluoroaniline hydrobromide ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Fluorophenylammonium bromide, 4-Fluoroanilinium bromide, 4-Fluorophenylamine hydrobromide

F1272-25G F1272-5G
TCIAF1272-25GEA 2590 DKK
TCIAF1272-25G TCIAF1272-5G
4-Fluoroaniline hydrobromide ≥98.0% (by HPLC, titration analysis)
4-Fluoroaniline hydrobromide
Formel: C₆H₆FN·HBr
CAS-nummer: 85734-18-5

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR