Fmoc-L-β-homoleucine ≥96.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Fmoc-L-β-HoLeu-OH, (S)-3-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-5-methylhexanoic acid, Fmoc-L-beta-homoleucine, Fmoc-β-HomoLeu-OH, Fmoc-beta-Homoleu-OH, [(9H-Fluoren-9-ylmethoxy)carbonyl]-L-beta-homoleucine

F1268-1G F1268-250MG
TCIAF1268-1GEA 1580 DKK
TCIAF1268-1G TCIAF1268-250MG
Fmoc-L-β-homoleucine ≥96.0% (by HPLC, titration analysis)
Fmoc-L-beta-homoleucine
Formel: C₂₂H₂₅NO₄
Smeltepunkt: 110°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01863059
CAS-nummer: 193887-44-4

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %
Specific rotation [a]20/D -26.0 to -31.0 deg (C=0.15, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR