2-Fluoroethylamine hydrochloride ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Aminoethyl Fluoride hydrochloride, 2-Fluoroethanamine hydrochloride, 2-Fluoroethylammonium hydrochloride

F1250-1G F1250-5G
TCIAF1250-1GEA 296 DKK
TCIAF1250-1G TCIAF1250-5G
2-Fluoroethylamine hydrochloride ≥97.0% (by titrimetric analysis)
2-Fluoroethylamine hydrochloride
Formel: C₂H₆FN·HCl
Molekylvægt: 99.53 g/mol
Smeltepunkt: 100°C
MDL Number: MFCD00008161
CAS-nummer: 460-08-2
EINECS: 207-302-3

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Water max. 3.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR