Fluphenazine dihydrochloride ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

F1240-1G F1240-5G
TCIAF1240-1GEA 846 DKK
TCIAF1240-1G TCIAF1240-5G
Fluphenazine dihydrochloride ≥98.0% (by HPLC)
Fluphenazine dihydrochloride
Formel: C₂₂H₂₈Cl₂F₃N₃OS
MDL Number: MFCD00055212
CAS-nummer: 146-56-5
EINECS: 205-674-1
UN nummer: 2811
ADR: 6.1,III
Merck Index: 14,04189

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 225.0 to 229.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR