2-Fluor-6-methoxybenzoesyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
F1146-1G
F1146-5G
TCIAF1146-1GEA
181
DKK
TCIAF1146-1G
TCIAF1146-5G
2-Fluor-6-methoxybenzoesyre ≥98.0% (by GC, titration analysis)
2-Fluor-6-methoxybenzoesyre
Formel:
C₈H₇FO₃ |
MDL Number:
MFCD00671764 CAS-nummer: 137654-21-8 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 90.0 to 94.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...