Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-tetradecanoyl-L-lysine ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Fmoc-Lys(myristoyl)-OH, Fmoc-Lys(Myr)-OH, Nalpha-Fmoc-Nepsilon-tetradecanoyl-L-lysine
F1143-1G
F1143-5G
TCIAF1143-1GEA
739
DKK
TCIAF1143-1G
TCIAF1143-5G
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-tetradecanoyl-L-lysine ≥98.0% (by HPLC, total nitrogen)
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-tetradecanoyl-L-lysine
Formel:
C₃₅H₅₀N₂O₅ |
CAS-nummer:
1128181-23-6 |
|
Specifikationer Testresultater
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 138.0 to 142.0 deg-C |
Specific rotation [a]20/D | +13.0 to +17.0 deg (C=1, CHCL3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...