4-Fluorophenyl-trans,trans-4'-n-pentylbicyclohexyl-4-carboxylate ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
trans,trans-4'-Pentylbicyclohexyl-4-carboxylic acid 4-Fluorophenyl ester, 4-Fluorophenyl trans,trans-4'-Pentylbicyclohexyl-4-carboxylate, (Trans,trans)-4-fluorophenyl 4'-pentyl-[1,1'-bi(cyclohexane)]-4-carboxylate
F1120-1G
F1120-5G
TCIAF1120-1GEA
370
DKK
TCIAF1120-1G
TCIAF1120-5G
4-Fluorophenyl-trans,trans-4'-n-pentylbicyclohexyl-4-carboxylate ≥98.0% (by GC)
4-Fluorophenyl-trans,trans-4'-n-pentylbicyclohexyl-4-carboxylate
Formel:
C₂₄H₃₅FO₂ |
MDL Number:
MFCD11053423 CAS-nummer: 88878-50-6 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...