4-Fluor-3-methylbenzoesyrechlorid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
F1116-5G
F1116-25G
TCIAF1116-5GEA
329
DKK
TCIAF1116-5G
TCIAF1116-25G
4-Fluor-3-methylbenzoesyrechlorid ≥98.0% (by GC, titration analysis)
4-Fluor-3-methylbenzoesyrechlorid
Formel:
C₈H₆ClFO Kogepunkt: 115°C Flammepunkt: 98°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD01631426 CAS-nummer: 455-84-5 UN nummer: 3265 ADR: 8,II |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...