6-Methoxy-4-oxo-4H-chromene-3-carbaldehyde ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 6-Methoxychromone-3-carboxaldehyde, 3-Formyl-6-methoxychromone

F1115-200MG F1115-1G
TCIAF1115-200MGEA 517 DKK
TCIAF1115-200MG TCIAF1115-1G
6-Methoxy-4-oxo-4H-chromene-3-carbaldehyde ≥98.0% (by GC)
6-Methoxy-4-oxo-4H-chromene-3-carbaldehyde
Formel: C₁₁H₈O₄
MDL Number: MFCD02683011
CAS-nummer: 42059-79-0

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 163.0 to 167.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR