trans,trans-4-(4-Fluorophenyl)-4'-propylbicyclohexyl ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F1102-1G
F1102-5G
TCIAF1102-1GEA
247
DKK
TCIAF1102-1G
TCIAF1102-5G
trans,trans-4-(4-Fluorophenyl)-4'-propylbicyclohexyl ≥98.0% (by GC)
trans,trans-4-(4-Fluorophenyl)-4'-propylbicyclohexyl
Formel:
C₂₁H₃₁F Smeltepunkt: 71°C |
MDL Number:
MFCD11053420 CAS-nummer: 82832-27-7 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Mesomorphic range | 85.0 to 160.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...