4'-Fluor-3'-methylacetophenon ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-(4-Fluoro-3-methylphenyl)ethanone

F1097-1G F1097-5G
TCIAF1097-1GEA 214 DKK
TCIAF1097-1G TCIAF1097-5G
4'-Fluor-3'-methylacetophenon ≥98.0% (by GC)
4'-Fluor-3'-methylacetophenon
Formel: C₉H₉FO
Molekylvægt: 152.17 g/mol
Kogepunkt: 215°C
MDL Number: MFCD04972879
CAS-nummer: 369-32-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d00b2d8c524e1885b3a331ad7718bd0f08cdc90d&vid=d00b2d8c524e1885b3a331ad7718bd0f08cdc90d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR