1-[(4-Fluorophenyl)carbamoyl]cyclopropanecarboxylic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

F1083-1G F1083-5G
TCIAF1083-1GEA 427 DKK
TCIAF1083-1G TCIAF1083-5G
1-[(4-Fluorophenyl)carbamoyl]cyclopropanecarboxylic acid ≥98.0% (by HPLC, titration analysis)
Formel: C₁₁H₁₀FNO₃
MDL Number: MFCD11226313
CAS-nummer: 849217-48-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7137d633b34dbaf8e687d7fa8f1b2ebb5eef4f25&vid=7137d633b34dbaf8e687d7fa8f1b2ebb5eef4f25&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 173.0 to 177.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR