2-Fluor-5-nitro-6-picolin ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 6-Fluoro-2-methyl-3-nitropyridine, 2-Fluoro-6-methyl-5-nitropyridine

F1067-1G F1067-5G
TCIAF1067-1GEA 189 DKK
TCIAF1067-1G TCIAF1067-5G
2-Fluor-5-nitro-6-picolin ≥97.0% (by GC)
2-Fluor-5-nitro-6-picolin
Formel: C₆H₅FN₂O₂
Molekylvægt: 156.12 g/mol
Kogepunkt: 85°C
MDL Number: MFCD03095071
CAS-nummer: 18605-16-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06d53f89d3384f964717191065d121a1fd9b0229&vid=06d53f89d3384f964717191065d121a1fd9b0229&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR