4-Fluoro-2-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Fluoro-2-(trifluoromethyl)benzeneboronic acid, (4-Fluoro-2-(trifluoromethyl)phenyl)boronic acid
F1052-1G
F1052-5G
TCIAF1052-1GEA
288
DKK
TCIAF1052-1G
TCIAF1052-5G
4-Fluoro-2-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
4-Fluoro-2-(trifluoromethyl)phenylboronic acid
Formel:
C₇H₅BF₄O₂ Molekylvægt: 207.92 g/mol |
MDL Number:
MFCD08062379 CAS-nummer: 182344-16-7 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...