5-(5-Fluoro-2-hydroxyphenyl)isoxazole ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Fluoro-2-(5-isoxazolyl)phenol, 5-(5-Fluoro-2-hydroxyphenyl)isoxazole

F1049-1G F1049-5G
TCIAF1049-1GEA 337 DKK
TCIAF1049-1G TCIAF1049-5G
5-(5-Fluoro-2-hydroxyphenyl)isoxazole ≥98.0% (by GC)
5-(5-Fluoro-2-hydroxyphenyl)isoxazole
Formel: C₉H₆FNO₂
MDL Number: MFCD03070563
CAS-nummer: 288401-62-7

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 188.0 to 192.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR