Furan-2,5-dicarbonyl dichloride ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2,5-Furandicarbonyl dichloride

F1022-1G F1022-5G
TCIAF1022-1GEA 583 DKK
TCIAF1022-1G TCIAF1022-5G
Furan-2,5-dicarbonyl dichloride ≥98.0% (by GC)
Furan-2,5-dicarbonyl dichloride
Formel: C₆H₂Cl₂O₃
Kogepunkt: 245°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD28044847
CAS-nummer: 10375-34-5
UN nummer: 3261
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 97.0 %
Melting point 76.0 to 80.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR