2-Fluoro-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Fluoro-3-(trifluoromethyl)benzeneboronic acid

F1006-1G F1006-5G
TCIAF1006-1GEA 214 DKK
TCIAF1006-1G TCIAF1006-5G
2-Fluoro-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
2-Fluoro-3-(trifluoromethyl)phenylboronic acid
Formel: C₇H₅BF₄O₂
MDL Number: MFCD07784336
CAS-nummer: 157834-21-4

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 95.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR