2-Fluor-5-iodtoluen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Fluoro-4-iodo-2-methylbenzene, 2-Fluoro-5-iodo-1-methylbenzene

F1004-5G F1004-25G
TCIAF1004-5GEA 312 DKK
TCIAF1004-5G TCIAF1004-25G
2-Fluor-5-iodtoluen ≥98.0% (by GC)
2-Fluor-5-iodtoluen
Formel: C₇H₆FI
Kogepunkt: 87°C
MDL Number: MFCD00013710
CAS-nummer: 452-68-6
EINECS: 207-206-1

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR