2-Fluor-4-trifluormethylpyridin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

F0995-1G F0995-5G
TCIAF0995-1GEA 435 DKK
TCIAF0995-1G TCIAF0995-5G
2-Fluor-4-trifluormethylpyridin ≥98.0% (by GC)
2-Fluor-4-trifluormethylpyridin
Formel: C₆H₃F₄N
Molekylvægt: 165.09 g/mol
Flammepunkt: 23°C
MDL Number: MFCD09879262
CAS-nummer: 118078-66-3
UN nummer: 1992
ADR: 3 III

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR