4-Fluorobenzil ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Fluorobibenzoyl, 1-(4-Fluorophenyl)-2-phenylethane-1,2-dione, 4-Fluorodibenzoyl

F0989-200MG F0989-1G
TCIAF0989-200MGEA 772 DKK
TCIAF0989-200MG TCIAF0989-1G
4-Fluorobenzil ≥98.0% (by GC)
4-Fluorobenzil
Formel: C₁₄H₉FO₂
MDL Number: MFCD00223483
CAS-nummer: 3834-66-0

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 66.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR