2-Fluor-3-nitropyridin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
F0982-5G
F0982-25G
TCIAF0982-5GEA
345
DKK
TCIAF0982-5G
TCIAF0982-25G
2-Fluor-3-nitropyridin ≥98.0% (by GC)
2-Fluoro-3-nitropyridine
Formel:
C₅H₃FN₂O₂ Kogepunkt: 110°C Smeltepunkt: 18°C |
MDL Number:
MFCD03095068 CAS-nummer: 1480-87-1 |
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...