N-(2-Fluorophenyl)-3-oxobutyramide ≥98.0% (by total nitrogen basis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N-(2-Fluorophenyl)-3-oxobutanamide, 2'-Fluoroacetoacetanilide

F0963-200MG F0963-1G
TCIAF0963-200MGEA 690 DKK
TCIAF0963-200MG TCIAF0963-1G
N-(2-Fluorophenyl)-3-oxobutyramide ≥98.0% (by total nitrogen basis)
N-(2-Fluorophenyl)-3-oxobutyramide
Formel: C₁₀H₁₀FNO₂
MDL Number: MFCD00661533
CAS-nummer: 5279-85-6

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 72.0 to 76.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR