4-Fluor-o-toluonitril ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

F0962-1G F0962-5G
TCIAF0962-1GEA 148 DKK
TCIAF0962-1G TCIAF0962-5G
4-Fluor-o-toluonitril ≥99.0% (by GC)
4-Fluor-o-toluonitril
Formel: C₈H₆FN
Molekylvægt: 135.14 g/mol
MDL Number: MFCD03095106
CAS-nummer: 147754-12-9
UN nummer: 3439
ADR: 6.1 III

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR