4-[[(4-Fluorophenyl)imino]methyl]phenol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: ALPHA-(4-FLUOROPHENYLIMINO)-P-CRESOL, 4-[[(4-Fluorophenyl)imino]methyl]phenol

F0946-5G F0946-25G
TCIAF0946-5GEA 444 DKK
TCIAF0946-5G TCIAF0946-25G
4-[[(4-Fluorophenyl)imino]methyl]phenol ≥98.0% (by GC, titration analysis)
4-[[(4-Fluorophenyl)imino]methyl]phenol
Formel: C₁₃H₁₀FNO
MDL Number: MFCD00029739
CAS-nummer: 3382-63-6

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 179.0 to 183.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR