4-(3-Fluorobenzyloxy)benzaldehyde ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
4-((3-Fluorobenzyl)oxy)benzaldehyde
F0943-1G
F0943-5G
TCIAF0943-1GEA
567
DKK
TCIAF0943-1G
TCIAF0943-5G
4-(3-Fluorobenzyloxy)benzaldehyde ≥98.0% (by GC)
4-(3-Fluorobenzyloxy)benzaldehyde
Formel:
C₁₄H₁₁FO₂ Molekylvægt: 230.24 g/mol Kogepunkt: 265°C |
MDL Number:
MFCD02605317 CAS-nummer: 66742-57-2 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 46.0 to 50.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...