(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-2-carboxylic acid, Fmoc-Pip-OH, 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-pipecolic Acid, Fmoc-HoPro-OH, 1-Fmoc-L-pipecolic Acid, Fmoc-HomoPro-OH, (S)-1-Fmoc-2-piperidinecarboxylic Acid
F0938-5G
F0938-1G
TCIAF0938-5GEA
1170
DKK
TCIAF0938-5G
TCIAF0938-1G
(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid ≥98.0% (by HPLC, titration analysis)
(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid
Formel:
C₂₁H₂₁NO₄ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00235898 CAS-nummer: 86069-86-5 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 155.0 to 159.0 deg-C |
Specific rotation [a]20/D | -20.0 to -24.0 deg (C=1,MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...