3-Fluorphenethylamin ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
F0930-5G
F0930-25G
TCIAF0930-5GEA
444
DKK
TCIAF0930-5G
TCIAF0930-25G
3-Fluorphenethylamin ≥98.0% (by GC, titration analysis)
3-Fluorphenethylamin
Formel:
C₈H₁₀FN Kogepunkt: 87°C Flammepunkt: 83°C |
MDL Number:
MFCD00075376 CAS-nummer: 404-70-6 UN nummer: 2735 ADR: 8,II |
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...