2-Furaldehyde diethyl acetal ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
2-(Diethoxymethyl)furan, Furfural Diethyl Acetal
F0929-5G
F0929-25G
TCIAF0929-5GEA
534
DKK
TCIAF0929-5G
TCIAF0929-25G
2-Furaldehyde diethyl acetal ≥98.0% (by GC)
2-Furaldehyde diethyl acetal
Formel:
C₉H₁₄O₃ Molekylvægt: 170.21 g/mol Kogepunkt: 81°C Flammepunkt: 64°C |
MDL Number:
MFCD00003245 CAS-nummer: 13529-27-6 |
Specifikationer Testresultater
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...