Fmoc-GABA-OH ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
4-(Fmoc-amino)butyric acid, N-Fmoc-4-aminobutyric acid, Fmoc-γ-Abu-OH, Fmoc-GABA-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-aminobutyric acid
F0911-5G
F0911-25G
TCIAF0911-5GEA
370
DKK
TCIAF0911-5G
TCIAF0911-25G
Fmoc-GABA-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-GABA-OH
Formel:
C₁₉H₁₉NO₄ Molekylvægt: 325.364 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00144889 CAS-nummer: 116821-47-7 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 172.0 to 176.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...