5-Fluoro-8-methoxyquinoline ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

F0909-1G F0909-5G
TCIAF0909-1GEA 542 DKK
TCIAF0909-1G TCIAF0909-5G
5-Fluoro-8-methoxyquinoline ≥98.0% (by GC, titration analysis)
5-Fluoro-8-methoxyquinoline
Formel: C₁₀H₈FNO
Molekylvægt: 177.18 g/mol
Kogepunkt: 147°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD18452297
CAS-nummer: 439-88-3

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR