4-Fluoro-N-salicylideneaniline ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-(((4-Fluorophenyl)imino)methyl)phenol, 4-Fluoro-N-salicylalaniline

F0893-1G F0893-5G
TCIAF0893-1GEA 238 DKK
TCIAF0893-1G TCIAF0893-5G
4-Fluoro-N-salicylideneaniline ≥98.0% (by GC, titration analysis)
4-Fluoro-N-salicylideneaniline
Formel: C₁₃H₁₀FNO
Molekylvægt: 215.23 g/mol
MDL Number: MFCD00020090
CAS-nummer: 3382-62-5

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 82.0 to 86.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR